• USA Home
  • Search Results

  • 246trihydroxyacetophenonemonohydrate1861624927828211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2′,4′,6′-Trihydroxyacetophenone monohydrate

2 Products
Synonym:   2-Acetylphloroglucinol, THAP
Image of 2′,4′,6′-Trihydroxyacetophenone monohydrate
CAS Number:
Linear Formula:
 (HO)3C6H2COCH3·H2O
Molecular Weight:
 186.16
 Beilstein/REAXYS Number:
 9288656
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More