• USA Home
  • Search Results

  • 246triiodophenol4718060923411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,4,6-Triiodophenol

2 Products
Image of 2,4,6-Triiodophenol
CAS Number:
Linear Formula:
 I3C6H2OH
Molecular Weight:
 471.80
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More