• USA Home
  • Search Results

  • 246tris2pyridylstriazine31233368235711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,4,6-Tris(2-pyridyl)-s-triazine

2 Products
Synonym:   2,4,6-Tri(2-pyridyl)-s-triazine, TPTZ
Image of 2,4,6-<WBR>Tris(2-<WBR>pyridyl)<WBR>-<I>s</I>-triazine
CAS Number:
Empirical Formula (Hill Notation):
 C18H12N6
Molecular Weight:
 312.33
 Beilstein/REAXYS Number:
 282581
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More