• USA Home
  • Search Results

  • 24d221049475711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,4-D

3 Products
Synonym:   2,4-Dichlorophenoxyacetic acid, 2,4-D
Image of 2,4-D
CAS Number:
Linear Formula:
 Cl2C6H3OCH2CO2H
Molecular Weight:
 221.04
 Beilstein/REAXYS Number:
 1214242
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Shelf Life
  Product Line
  Anal. Standard Form
  Physical Form
  Special Grade
Show More