• USA Home
  • Search Results

  • 24diamino6hydroxypyrimidine126125606411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,4-Diamino-6-hydroxypyrimidine

1 Products
Synonym:   2,4-Diamino-6-hydroxypyrimidine, 2,6-Diamino-4-pyrimidinol, 2,6-Diamino-4-pyrimidinone
Image of 2,4-<WBR>Diamino-<WBR>6-<WBR>hydroxypyrimidine
CAS Number:
Empirical Formula (Hill Notation):
 C4H6N4O
Molecular Weight:
 126.12
 Beilstein/REAXYS Number:
 125006
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More