• USA Home
  • Search Results

  • 24dibromoaniline2509261557611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,4-Dibromoaniline

1 Products
Image of 2,4-Dibromoaniline
CAS Number:
Linear Formula:
 Br2C6H3NH2
Molecular Weight:
 250.92
 Beilstein/REAXYS Number:
 2206653
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
D38405 2,4-Dibromoaniline, 98%,