• USA Home
  • Search Results

  • 24dibromophenol2519061558711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,4-Dibromophenol

1 Products
Image of 2,4-Dibromophenol
CAS Number:
Linear Formula:
 Br2C6H3OH
Molecular Weight:
 251.90
 Beilstein/REAXYS Number:
 1861291
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More

Product #
Description
258164 2,4-Dibromophenol, 95%,