• USA Home
  • Search Results

  • 24dichlorobenzylalcohol17703177782811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,4-Dichlorobenzyl alcohol

2 Products
Image of 2,4-Dichlorobenzyl alcohol
CAS Number:
Linear Formula:
 Cl2C6H3CH2OH
Molecular Weight:
 177.03
 Beilstein/REAXYS Number:
 1448652
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More