• USA Home
  • Search Results

  • 24dimethyl3pentanol1162060036211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,4-Dimethyl-3-pentanol

1 Products
Synonym:   Diisopropylcarbinol
Image of 2,4-<WBR>Dimethyl-<WBR>3-<WBR>pentanol
CAS Number:
Linear Formula:
 (CH3)2CHCH(OH)CH(CH3)2
Molecular Weight:
 116.20
 Beilstein/REAXYS Number:
 1731593
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More