• USA Home
  • Search Results

  • 24dimethylaminostyryl1ethylpyridiniumiodide38027378501111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-[4-(Dimethylamino)styryl]-1-ethylpyridinium iodide

1 Products
Synonym:   DASPEI
Image of 2-<WBR>[4-<WBR>(Dimethylamino)<WBR>styryl]<WBR>-<WBR>1-<WBR>ethylpyridinium iodide
CAS Number:
Empirical Formula (Hill Notation):
 C17H21IN2
Molecular Weight:
 380.27
 Beilstein/REAXYS Number:
 3783243
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More