• USA Home
  • Search Results

  • 24dimethylphenol1221610567911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,4-Dimethylphenol

2 Products
Synonym:   2,4-DMP, asym.-m-Xylenol, 2,4-DMP, asym.-m-Xylenol
Image of 2,4-Dimethylphenol
CAS Number:
Linear Formula:
 2,4-(CH3)2C6H3OH
Molecular Weight:
 122.16
 EC Index Number
 203-321-6
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Anal. Standard Form
  Physical Form
  Special Grade
Show More