• USA Home
  • Search Results

  • 24dinitrotoluene1821312114211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,4-Dinitrotoluene

3 Products
Image of 2,4-Dinitrotoluene
CAS Number:
Linear Formula:
 CH3C6H3(NO2)2
Molecular Weight:
 182.13
 Beilstein/REAXYS Number:
 1912834
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Shelf Life
  Special Grade
Show More