• USA Home
  • Search Results

  • 24dinitrotoluenesolution1821312114211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,4-Dinitrotoluene solution

2 Products
Image of 2,4-Dinitrotoluene solution
CAS Number:
Empirical Formula (Hill Notation):
 C7H6N2O4
Molecular Weight:
 182.13
 MDL number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Anal. Standard Form
  Special Grade
  Package quantity
  Manufacturer designation
  Feature
Show More