• USA Home
  • Search Results

  • 24ditertbutyl65chloro2hbenzotriazol2ylphenol35788386499111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,4-Di-tert-butyl-6-(5-chloro-2H-benzotriazol-2-yl)phenol

2 Products
Synonym:   2,4-Di-(tert -butyl)-6-(5-chloro-2H -benzo[d ][1,2,3]triazol-2-yl)phenol, 2-(2-Hydroxy-3,5-di-tert -butylphenyl)-5-chloro-2H -benzotriazole, 2-(2-Hydroxy-3,5-di-tert -butylphenyl)-5-chlorobenzotriazole, 2-(5-Chloro-2H -benzotriazol-2-yl)-4,6-bis(1,1-dimethylethyl)phenol, 5-Chloro-2-(3,5-di-tert -butyl-2-hydroxyphenyl)benzotriazole
Image of 2,4-Di-<I>tert</I>-butyl-6-(5-chloro-2<I>H</I>-<WBR>benzotriazol-<WBR>2-<WBR>yl)<WBR>phenol
CAS Number:
Empirical Formula (Hill Notation):
 C20H24ClN3O
Molecular Weight:
 357.88
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More