• USA Home
  • Search Results

  • 24dsolution123459475711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,4-D solution

1 Products
Synonym:   2,4-Dichlorophenoxyacetic acid solution, 2,4-D
CAS Number:
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Anal. Standard Form
  Special Grade
  Application(s)
  Package quantity
Show More