• USA Home
  • Search Results

  • 24hydroxyphenylazobenzoicacid24223163482811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-(4-Hydroxyphenylazo)benzoic acid

2 Products
Synonym:   2-(4′-Hydroxybenzeneazo)benzoic acid, 4′-Hydroxyazobenzene-2-carboxylic acid, HABA, HBABA
Image of 2-<WBR>(4-<WBR>Hydroxyphenylazo)<WBR>benzoic acid
CAS Number:
Linear Formula:
 HOC6H4N=NC6H4CO2H
Molecular Weight:
 242.23
 Beilstein/REAXYS Number:
 1842696
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Absorption
  Special Grade
  Melting Point
  Solubility
Show More