• USA Home
  • Search Results

  • 24pyridinedicarboxylicacid1671249980911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,4-Pyridinedicarboxylic acid

2 Products
Synonym:   Lutidinic acid
Image of 2,4-<WBR>Pyridinedicarboxylic acid
CAS Number:
Empirical Formula (Hill Notation):
 C7H5NO4
Molecular Weight:
 167.12
 Beilstein/REAXYS Number:
 131631
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More