• USA Home
  • Search Results

  • 24thiazolidinedione11713229531011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,4-Thiazolidinedione

1 Products
Image of 2,4-<WBR>Thiazolidinedione
CAS Number:
Empirical Formula (Hill Notation):
 C3H3NO2S
Molecular Weight:
 117.13
 Beilstein Registry Number:
 110700
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More