• USA Home
  • Search Results

  • 25dichlorobenzotrifluoride2150032050311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,5-Dichlorobenzotrifluoride

1 Products
Synonym:   2,5-Dichloro-α,α,α-trifluorotoluene
Image of 2,5-<WBR>Dichlorobenzotrifluoride
CAS Number:
Linear Formula:
 Cl2C6H3CF3
Molecular Weight:
 215.00
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More