• USA Home
  • Search Results

  • 25dichlorobenzylalcohol177033414505611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,5-Dichlorobenzyl alcohol

1 Products
Image of 2,5-Dichlorobenzyl alcohol
CAS Number:
Linear Formula:
 Cl2C6H3CH2OH
Molecular Weight:
 177.03
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More