• USA Home
  • Search Results

  • 25difluorophenol13009271331711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,5-Difluorophenol

1 Products
Image of 2,5-Difluorophenol
CAS Number:
Linear Formula:
 F2C6H3OH
Molecular Weight:
 130.09
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
290394 2,5-Difluorophenol, 95%,