• USA Home
  • Search Results

  • 25dihydrofuran7009170829811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,5-Dihydrofuran

1 Products
Image of 2,5-Dihydrofuran
CAS Number:
Empirical Formula (Hill Notation):
 C4H6O
Molecular Weight:
 70.09
 Beilstein/REAXYS Number:
 103174
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
253170 2,5-Dihydrofuran, 97%,