• USA Home
  • Search Results

  • 25dihydroxybenzoicacid1541249079911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,5-Dihydroxybenzoic acid

5 Products
Synonym:   2,5-DHBA, DHB, Gentisic acid, Hydroquinonecarboxylic acid
Image of 2,5-Dihydroxybenzoic acid
CAS Number:
Linear Formula:
 (HO)2C6H3CO2H
Molecular Weight:
 154.12
 Beilstein/REAXYS Number:
 2209119
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Analyte Suitability
  Special Grade
  Melting Point
  Application(s)
Show More