• USA Home
  • Search Results

  • 25dimethylbenzenethiol13823400161011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,5-Dimethylbenzenethiol

1 Products
Synonym:   2,5-Dimethylthiophenol
Image of 2,5-<WBR>Dimethylbenzenethiol
CAS Number:
Linear Formula:
 (CH3)2C6H3SH
Molecular Weight:
 138.23
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More