• USA Home
  • Search Results

  • 25dimethylcelecoxib3954045763926811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,5-Dimethyl-celecoxib

2 Products
Synonym:   2,5-Dimethylcelecoxib, 4-[5-(2,5-dimethylphenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]-benzenesulfonamide
Image of 2,5-<WBR>Dimethyl-<WBR>celecoxib
CAS Number:
Empirical Formula (Hill Notation):
 C18H16F3N3O2S
Molecular Weight:
 395.40
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Physical Form
Show More