• USA Home
  • Search Results

  • 25dimethylphenol122169587411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,5-Dimethylphenol

3 Products
Synonym:   2-Hydroxy-p-xylene, p-Xylenol
Image of 2,5-Dimethylphenol
CAS Number:
Linear Formula:
 (CH3)2C6H3OH
Molecular Weight:
 122.16
 Beilstein/REAXYS Number:
 1099260
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Physical Form
  Special Grade
  Melting Point
Show More