• USA Home
  • Search Results

  • 25ditertbutylhydroquinone222328858411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,5-Di-tert-butylhydroquinone

2 Products
Image of 2,5-Di-<I>tert</I>-butylhydroquinone
CAS Number:
Linear Formula:
 [(CH3)3C]2C6H2-1,4-(OH)2
Molecular Weight:
 222.32
 Beilstein/REAXYS Number:
 2049542
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More