• USA Home
  • Search Results

  • 25hydroxycholecalciferol400641935617311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

25-Hydroxycholecalciferol

1 Products
Synonym:   25-Hydroxyvitamin D3
Image of 25-<WBR>Hydroxycholecalciferol
CAS Number:
Empirical Formula (Hill Notation):
 C27H44O2
Molecular Weight:
 400.64
 EC Number:
 MDL number:
 NACRES:
 NA.79
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More