• USA Home
  • Search Results

  • 25hydroxyvitamind261919d3solution41567121746739411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

25-Hydroxyvitamin D2 (6,19,19-d3) solution

3 Products
Synonym:   25-Hydroxyergocalciferol (6,19,19-d3) solution, 25-Hydroxycalciferol (6,19,19-d3) solution
Image of 25-Hydroxyvitamin D<SUB>2</SUB> (6,19,19-d<SUB>3</SUB>) solution
CAS Number:
Empirical Formula (Hill Notation):
 C28D3H41O2
Molecular Weight:
 415.67
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Melting Point
  Physical Form
  Isotopic Purity
  Titertest
  Concentration
  Color
Show More