• USA Home
  • Search Results

  • 25hydroxyvitamind3monohydrate418656328336311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

25-Hydroxyvitamin D3 monohydrate

1 Products
Synonym:   25-Hydroxycholecalciferol
Image of 25-Hydroxyvitamin D<SUB>3</SUB> monohydrate
CAS Number:
Empirical Formula (Hill Notation):
 C27H44O2 · H2O
Molecular Weight:
 418.65
 Beilstein/REAXYS Number:
 4270041
 MDL number:
 NACRES:
 NA.79
 PubChem Substance ID:
 eCl@ss
 34058003
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More