• USA Home
  • Search Results

  • 25hydroxyvitamind3solution400641935617311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

25-Hydroxyvitamin D3 solution

3 Products
Synonym:   25-Hydroxycholecalciferol solution
Image of 25-Hydroxyvitamin D<SUB>3</SUB> solution
CAS Number:
Empirical Formula (Hill Notation):
 C27H44O2
Molecular Weight:
 400.64
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
  Concentration
  Color
  Assay
Show More