• USA Home
  • Search Results

  • 25xylenol122169587411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,5-Xylenol

1 Products
Synonym:   2,5-Dimethylphenol, 2-Hydroxy-p-xylene, p-Xylenol
Image of 2,5-Xylenol
CAS Number:
Linear Formula:
 (CH3)2C6H3OH
Molecular Weight:
 122.16
 Beilstein/REAXYS Number:
 1099260
 Council of Europe no.:
 537
 EC Number:
 FEMA Number:
 3595
 Flavis number:
 4.019
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More

Product #
Description
W359505 2,5-Xylenol, ≥99%, FG,