• USA Home
  • Search Results

  • 26dichloroindophenolsodiumsalthydrate29008anhydrousbasis126661556811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,6-Dichloroindophenol sodium salt hydrate

1 Products
Synonym:   2,6-Dichlorobenzenone-indophenol sodium salt, 2,6-Dichlorophenolindophenol sodium salt hydrate, 2,6-Dichloro-N-(4-hydroxyphenyl)-1,4-benzoquinoneimine sodium salt, DCIP, DPIP
Image of 2,6-<WBR>Dichloroindophenol sodium salt hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C12H6Cl2NNaO2 · xH2O
Molecular Weight:
 290.08 (anhydrous basis)
 Beilstein/REAXYS Number:
 3641229
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More