• USA Home
  • Search Results

  • 26dichloropyridine3carbonitrile173004038190611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,6-Dichloropyridine-3-carbonitrile

2 Products
Synonym:   2,6-Dichloronicotinonitrile
Image of 2,6-<WBR>Dichloropyridine-<WBR>3-<WBR>carbonitrile
CAS Number:
Empirical Formula (Hill Notation):
 C6H2Cl2N2
Molecular Weight:
 173.00
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More