• USA Home
  • Search Results

  • 26difluorophenol130092817748211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,6-Difluorophenol

1 Products
Image of 2,6-Difluorophenol
CAS Number:
Linear Formula:
 F2C6H3OH
Molecular Weight:
 130.09
 Beilstein/REAXYS Number:
 2043613
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
264466 2,6-Difluorophenol, 98%,