• USA Home
  • Search Results

  • 26dihydroxyacetophenone1521569983211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2′,6′-Dihydroxyacetophenone

2 Products
Synonym:   2-Acetyl-1,3-dihydroxybenzene, 2-Acetylresorcinol, DHAP
Image of 2′,6′-Dihydroxyacetophenone
CAS Number:
Linear Formula:
 (HO)2C6H3COCH3
Molecular Weight:
 152.15
 Beilstein/REAXYS Number:
 1366061
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More