• USA Home
  • Search Results

  • 26diisopropylphenol17827207854811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,6-Diisopropylphenol

2 Products
Synonym:   2,6-Bis(1-methylethyl)phenol, 2,6-Bis(isopropyl)phenol, Propofol
Image of 2,6-<WBR>Diisopropylphenol
CAS Number:
Linear Formula:
 [(CH3)2CH]2C6H3OH
Molecular Weight:
 178.27
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
Show More