• USA Home
  • Search Results

  • 26diisopropylphenylimidoneophylidenesbiphenmolybdenumvi7559220581580111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,6-Diisopropylphenylimido-neophylidene[(S)-(−)-BIPHEN]molybdenum(VI)

1 Products
Synonym:   (S)-Schrock-Hoveyda Catalyst
Image of 2,6-<WBR>Diisopropylphenylimido-<WBR>neophylidene[(<I>S</I>)-(−)-BIPHEN]molybdenum(VI)
CAS Number:
Empirical Formula (Hill Notation):
 C46H61MoNO2
Molecular Weight:
 755.92
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More