• USA Home
  • Search Results

  • 26dimethoxyphenol154169110111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,6-Dimethoxyphenol

4 Products
Synonym:   Pyrogallol 1,3-dimethyl ether
Image of 2,6-Dimethoxyphenol
CAS Number:
Linear Formula:
 (CH3O)2C6H3OH
Molecular Weight:
 154.16
 Beilstein/REAXYS Number:
 1526871
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Titertest
  Melting Point
  Special Grade
  Assay
Show More