• USA Home
  • Search Results

  • 26dimethyl4heptanone1422410883811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,6-Dimethyl-4-heptanone

4 Products
Synonym:   Diisobutyl ketone, Isovalerone
Image of 2,6-<WBR>Dimethyl-<WBR>4-<WBR>heptanone
CAS Number:
Linear Formula:
 (CH3)2CHCH2COCH2CH(CH3)2
Molecular Weight:
 142.24
 Beilstein/REAXYS Number:
 1743163
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More