• USA Home
  • Search Results

  • 26dimethyl5heptenal1402210672911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,6-Dimethyl-5-heptenal

3 Products
Synonym:   Melonal
Image of 2,6-<WBR>Dimethyl-<WBR>5-<WBR>heptenal
CAS Number:
Linear Formula:
 (CH3)2C=CHCH2CH2CH(CH3)CHO
Molecular Weight:
 140.22
 Council of Europe no.:
 2006
 EC Number:
 FEMA Number:
 2389
 Flavis number:
 5.074
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Density
  Special Grade
Show More