• USA Home
  • Search Results

  • 26dimethylbenzoicacid1501763246211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,6-Dimethylbenzoic acid

1 Products
Synonym:   vic.-m-Xylylic acid
Image of 2,6-Dimethylbenzoic acid
CAS Number:
Linear Formula:
 (CH3)2C6H3CO2H
Molecular Weight:
 150.17
 Beilstein/REAXYS Number:
 1863791
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More