• USA Home
  • Search Results

  • 26dimethylphenylisocyanide13117276971311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,6-Dimethylphenyl isocyanide

1 Products
Synonym:   vic.-m-Xylyl isocyanide
Image of 2,6-Dimethylphenyl isocyanide
CAS Number:
Linear Formula:
 (CH3)2C6H3NC
Molecular Weight:
 131.17
 Beilstein/REAXYS Number:
 3541909
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Melting Point
Show More