• USA Home
  • Search Results

  • 26naphthalenedicarboxylicacid21619114138411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,6-Naphthalenedicarboxylic acid

2 Products
Image of 2,6-<WBR>Naphthalenedicarboxylic acid
CAS Number:
Linear Formula:
 C10H6(CO2H)2
Molecular Weight:
 216.19
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More