• USA Home
  • Search Results

  • 26pyridinedicarboxylicacid1671249983211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,6-Pyridinedicarboxylic acid

2 Products
Synonym:   DPA, DPAc, Dipicolinic acid
Image of 2,6-<WBR>Pyridinedicarboxylic acid
CAS Number:
Empirical Formula (Hill Notation):
 C7H5NO4
Molecular Weight:
 167.12
 Beilstein/REAXYS Number:
 131629
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss:
 39151816
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More