• USA Home
  • Search Results

  • 27diaminofluorene1962552564411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,7-Diaminofluorene

1 Products
Synonym:   2,7-Fluorenediamine
Image of 2,7-Diaminofluorene
CAS Number:
Empirical Formula (Hill Notation):
 C13H12N2
Molecular Weight:
 196.25
 Beilstein/REAXYS Number:
 2099859
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
D17106 2,7-Diaminofluorene, >97%,