• USA Home
  • Search Results

  • 27dichlorofluorescein401207654011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2′,7′-Dichlorofluorescein

4 Products
Image of 2′,7′-Dichlorofluorescein
CAS Number:
Empirical Formula (Hill Notation):
 C20H10Cl2O5
Molecular Weight:
 401.20
 Beilstein/REAXYS Number:
 58009
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More