• USA Home
  • Search Results

  • 27dimethylnaphthalene1562258216111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,7-Dimethylnaphthalene

1 Products
Image of 2,7-<WBR>Dimethylnaphthalene
CAS Number:
Linear Formula:
 C10H6(CH3)2
Molecular Weight:
 156.22
 Beilstein/REAXYS Number:
 1852737
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More