• USA Home
  • Search Results

  • 2acetamidophenol1511661480211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Acetamidophenol

1 Products
Synonym:   2′-Hydroxyacetanilide, N-(2-Hydroxyphenyl)acetamide, NSC 3989
Image of 2-Acetamidophenol
CAS Number:
Linear Formula:
 CH3CONHC6H4OH
Molecular Weight:
 151.16
 Beilstein/REAXYS Number:
 607592
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
A7000 2-Acetamidophenol, 97%,