• USA Home
  • Search Results

  • 2acetyl1methylpyrrole1231593216111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2-Acetyl-1-methylpyrrole

2 Products
Image of 2-<WBR>Acetyl-<WBR>1-<WBR>methylpyrrole
CAS Number:
Empirical Formula (Hill Notation):
 C7H9NO
Molecular Weight:
 123.15
 Council of Europe no.:
 11373
 EC Number:
 FEMA Number:
 3184
 Flavis number:
 14.046
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Assay
Show More